Gene Transcripts
- Gene Symbol
- Ly6f
- Entrez Gene ID
- 17071
- Gene Name
- lymphocyte antigen 6 complex, locus F
- Gene Aliases
- Chromosome Location
- Chr. 15: 75268372 - 75272236 on Build GRCm38
- UniGene ID
- Mm.377095
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008530.2 |
NP_032556.1
|
4 |
442 |
|
View Full Product Details