Gene Transcripts

Gene Symbol
Vmn1r19
Entrez Gene ID
171200
Gene Name
vomeronasal 1 receptor 19
Gene Aliases
V1rc27
Chromosome Location
Chr. 6: 57400820 - 57405477 on Build GRCm38
UniGene ID
Mm.377178
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134182.1 NP_598943.1 1 546
GenBank mRNA BC125351.1 - 1 644
BC132235.1 - 1 644 40130512

View Full Product Details