Gene Transcripts

Gene Symbol
Havcr1
Entrez Gene ID
171283
Gene Name
hepatitis A virus cellular receptor 1
Gene Aliases
AI503787, KIM-1, TIM-1, Tim1, Timd1
Chromosome Location
Chr. 11: 46739823 - 46779578 on Build GRCm38
UniGene ID
Mm.17771
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166631.1 NP_001160103.1 3 505
NM_001166632.1 NP_001160104.1 3 389
NM_134248.2 NP_599009.2 2 344
GenBank mRNA AF399829.1 - 2 335
AF399830.1 - 2 335
AK133466.1 - 3 507
BC053400.1 - 3 397 30033601
CA551363.1 - 3 389

View Full Product Details