Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008963.2 | NP_032989.2 | 4 | 511 | |
GenBank mRNA | AB006361.1 | - | 4 | 494 | |
AK002382.1 | - | 3 | 336 | ||
AK131859.1 | - | 4 | 511 | ||
BC038083.1 | - | 3 | 587 | 4481308 | |
BC043015.1 | - | 4 | 497 | 5683706 | |
X89222.1 | - | 4 | 482 |