Gene Transcripts

Gene Symbol
Ptprj
Entrez Gene ID
19271
Gene Name
protein tyrosine phosphatase, receptor type, J
Gene Aliases
AI450271, BET, Byp, CD148, DEP-1, PTPbeta2, Ptpb2, RPTPJ, Scc-1, Scc1
Chromosome Location
Chr. 2: 90429756 - 90580677 on Build GRCm38
UniGene ID
Mm.330393
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135657.1 NP_001129129.1 9 1830
NM_008982.5 NP_033008.3 9 2114
GenBank mRNA AK147318.1 - 9 2035
AK147556.1 - 9 2035
AY038891.1 - 9 1932
AY039232.1 - 9 1932
D45212.1 - 9 2070
D83203.1 - 9 1831
DQ133576.1 - 9 1845

View Full Product Details