Gene Transcripts

Gene Symbol
Gm12
Entrez Gene ID
192852
Gene Name
predicted gene 12
Gene Aliases
Chromosome Location
Chr. 11: 98598291 - 98599586 on Build GRCm38
UniGene ID
Mm.342821
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195544.1 NP_001182473.1 2 339
GenBank mRNA BY232969.1 - 2 339

View Full Product Details