Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009251.1 | NP_033277.1 | 1398 | ||
GenBank mRNA | AA210495.1 | - | 3 | 463 | |
AK156462.1 | - | 5 | 1396 | ||
AY862185.1 | - | 1358 | |||
BC002065.1 | - | 5 | 1187 | ||
BC057144.1 | - | 4 | 1039 | ||
BC096664.1 | - | 4 | 700 | ||
BC119537.2 | - | 1414 | 40056419 | ||
BE850123.1 | - | 1 | 62 | ||
M64085.1 | - | 6 | 1161 |