Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001177667.1 | NP_001171138.1 | 39 | 5194 | |
NM_001177668.1 | NP_001171139.1 | 38 | 5134 | ||
GenBank mRNA | AK147317.1 | - | 38 | 5020 | |
BC022120.1 | - | 8 | 887 | ||
BC027791.1 | - | 15 | 1611 | ||
BC150941.1 | - | 39 | 5030 | 9087852 | |
BC158015.1 | - | 39 | 5030 | 9007321 | |
BC172095.1 | - | 38 | 4970 | 9054381 |