Gene Transcripts
- Gene Symbol
- Duox2
- Entrez Gene ID
- 214593
- Gene Name
- dual oxidase 2
- Gene Aliases
- A430065P05Rik, LNOX2, NOXEF2, P138-TOX, THOX2
- Chromosome Location
- Chr. 2: 122279246 - 122298742 on Build GRCm38
- UniGene ID
- Mm.358846
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_177610.2 |
NP_808278.2
|
2 |
116 |
|
View Full Product Details