Gene Transcripts

Gene Symbol
Traf4
Entrez Gene ID
22032
Gene Name
TNF receptor associated factor 4
Gene Aliases
A530032M13Rik, CART1, msp2
Chromosome Location
Chr. 11: 78158423 - 78165569 on Build GRCm38
UniGene ID
Mm.390418
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009423.4 NP_033449.2 7 1033
GenBank mRNA AF233449.1 - 7 932
AK035056.1 - 7 1011
AK040877.1 - 1011
AK046015.1 - 6 1317
AK053900.1 - 7 1030
AK198624.1 - 1 117
AK205707.1 - 1 59
BC020455.1 - 7 1018

View Full Product Details