Gene Transcripts
- Gene Symbol
- Trpc4
- Entrez Gene ID
- 22066
- Gene Name
- transient receptor potential cation channel, subfamily C, member 4
- Gene Aliases
- CCE1, STRPC4, Trp4, Trrp4
- Chromosome Location
- Chr. 3: 54156057 - 54318471 on Build GRCm38
- UniGene ID
- Mm.10100
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001253682.1 |
NP_001240611.1
|
8, 9 |
2323 |
|
NM_016984.3 |
NP_058680.1
|
8, 9 |
2323 |
|
GenBank mRNA |
AF011543.1 |
- |
8, 9 |
2140 |
|
AF019663.1 |
- |
8, 9 |
2140 |
|
AF190646.1 |
- |
8, 9 |
2214 |
|
AK044265.1 |
- |
8, 9 |
2179 |
|
AK080670.1 |
- |
8, 9 |
2324 |
|
BC057335.1 |
- |
8, 9 |
2159 |
|
BC060160.1 |
- |
8, 9 |
2159 |
|
BC120729.1 |
- |
8, 9 |
2238 |
40129652 |
BC131915.1 |
- |
8, 9 |
2238 |
40130192 |
U50921.1 |
- |
8, 9 |
2276 |
|
U50922.1 |
- |
8, 9 |
2276 |
|
View Full Product Details