Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001168288.1 | NP_001161760.1 | 6 | 2479 | |
NM_198420.2 | NP_940812.2 | 6 | 2479 | ||
GenBank mRNA | AK035479.1 | - | 5 | 2184 | |
AK141623.1 | - | 6 | 2440 | ||
AK173232.1 | - | 6 | 2414 | ||
BC060065.1 | - | 2 | 1051 | ||
BC060637.1 | - | 6 | 2460 | 6849400 |