Gene Transcripts

Gene Symbol
Lgi4
Entrez Gene ID
243914
Gene Name
leucine-rich repeat LGI family, member 4
Gene Aliases
Lgil3, clp
Chromosome Location
Chr. 7: 31058953 - 31070935 on Build GRCm38
UniGene ID
Mm.389870
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144556.2 NP_653139.2 9 1622
GenBank mRNA AJ487521.1 - 9 1623
AJ487960.1 - 9 1622
AK033967.1 - 9 1440
AK134643.1 - 9 2216
AK148793.1 - 9 1882
BC098469.1 - 4 1118 30099294

Gene Symbol
Fxyd3
Entrez Gene ID
17178
Gene Name
FXYD domain-containing ion transport regulator 3
Gene Aliases
Mat8
Chromosome Location
Chr. 7: 31070535 - 31076697 on Build GRCm38
UniGene ID
Mm.263847
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK145206.1 - 1922

View Full Product Details