Gene Transcripts

Gene Symbol
Cdk8
Entrez Gene ID
264064
Gene Name
cyclin-dependent kinase 8
Gene Aliases
Chromosome Location
Chr. 5: 146230736 - 146302874 on Build GRCm38
UniGene ID
Mm.260576
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153599.3 NP_705827.2 7 779
GenBank mRNA AK131948.1 - 7 703
CF723869.1 - 5 481
CN665544.1 - 1 43

View Full Product Details