Gene Transcripts

Gene Symbol
Celf5
Entrez Gene ID
319586
Gene Name
CUGBP, Elav-like family member 5
Gene Aliases
4930565A21Rik, Brunol5
Chromosome Location
Chr. 10: 81459227 - 81482725 on Build GRCm38
UniGene ID
Mm.152689
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176954.3 NP_795928.2 9 1075
GenBank mRNA AK029796.1 - 9 2249
AK132321.1 - 9 1041
AK158669.1 - 8 852
BC079845.1 - 9 1046

View Full Product Details