Gene Transcripts

Gene Symbol
Fbxw21
Entrez Gene ID
320082
Gene Name
F-box and WD-40 domain protein 21
Gene Aliases
E330009P21Rik
Chromosome Location
Chr. 9: 109136887 - 109162022 on Build GRCm38
UniGene ID
Mm.188028
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177069.3 NP_796043.1 9 1219
GenBank mRNA AK087710.1 - 9 1220
AK135949.1 - 9 1220
AK136136.1 - 9 1216
BC099397.1 - 9 1201 30936381

View Full Product Details