Gene Transcripts

Gene Symbol
Samd5
Entrez Gene ID
320825
Gene Name
sterile alpha motif domain containing 5
Gene Aliases
E130306M17Rik
Chromosome Location
Chr. 10: 9627259 - 9675208 on Build GRCm38
UniGene ID
Mm.101115
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177271.3 NP_796245.2 1 269
GenBank mRNA AK087498.1 - 1 270
BC145672.1 - 1 43 40130975
BC145674.1 - 1 43 40130977

View Full Product Details