Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_181858.1 | NP_862906.1 | 188 | ||
GenBank mRNA | BC038128.1 | - | 224 | ||
BC059731.1 | - | 163 | |||
BC139203.1 | - | 4 | 235 | 8862225 | |
BC145912.1 | - | 4 | 235 | 40131215 | |
BC171969.1 | - | 4 | 235 | 9054255 |