Gene Transcripts

Gene Symbol
Txlnb
Entrez Gene ID
378431
Gene Name
taxilin beta
Gene Aliases
2310001N14Rik, Mdp77
Chromosome Location
Chr. 10: 17796091 - 17845665 on Build GRCm38
UniGene ID
Mm.283493
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138628.3 NP_619534.2 7 1251
GenBank mRNA AF422244.1 - 7 1188
AF422245.1 - 7 1188
AK084639.1 - 7 1252
AK085904.1 - 7 1147
AK137442.1 - 7 1166
BC141023.1 - 7 1164 9055635

View Full Product Details