Gene Transcripts
- Gene Symbol
- Kng2
- Entrez Gene ID
- 385643
- Gene Name
- kininogen 2
- Gene Aliases
- Chromosome Location
- Chr. 16: 22985852 - 23029101 on Build GRCm38
- UniGene ID
- Mm.440414
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001102409.1 |
NP_001095879.1
|
3, 4 |
571 |
|
NM_001102410.1 |
NP_001095880.1
|
3, 4 |
571 |
|
NM_201375.2 |
NP_958763.1
|
3, 4 |
571 |
|
GenBank mRNA |
AY462056.1 |
- |
3, 4 |
487 |
|
AY462057.1 |
- |
3, 4 |
487 |
|
AY462059.1 |
- |
3, 4 |
487 |
|
AY660572.1 |
- |
3, 4 |
487 |
|
BC108335.1 |
- |
3, 4 |
538 |
5124483 |
BC108934.1 |
- |
3, 4 |
479 |
|
BC139720.1 |
- |
3, 4 |
479 |
40039854 |
View Full Product Details