Gene Transcripts

Gene Symbol
Wfdc13
Entrez Gene ID
408190
Gene Name
WAP four-disulfide core domain 13
Gene Aliases
Wfdc13l1
Chromosome Location
Chr. 2: 164685107 - 164687706 on Build GRCm38
UniGene ID
Mm.376205
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012704.1 NP_001012722.1 1 178
GenBank mRNA AY542486.1 - 1 178
AY542487.1 - 1 178
BC145778.1 - 1 87 40131081
BC145780.1 - 1 87 40131083

View Full Product Details