Gene Transcripts
- Gene Symbol
- Tomm40
- Entrez Gene ID
- 53333
- Gene Name
- translocase of outer mitochondrial membrane 40 homolog (yeast)
- Gene Aliases
- AW539759, Mom35, Tom40
- Chromosome Location
- Chr. 7: 19701313 - 19715429 on Build GRCm38
- UniGene ID
- Mm.416870
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001109748.1 |
NP_001103218.1
|
6, 7 |
950 |
|
NM_016871.2 |
NP_058567.2
|
7, 8 |
852 |
|
GenBank mRNA |
AF043249.1 |
- |
6, 7 |
628 |
|
AF316403.1 |
- |
6, 7 |
785 |
|
AF316404.1 |
- |
6, 7 |
785 |
|
AK142575.1 |
- |
6, 7 |
546 |
|
AK150019.1 |
- |
6, 7 |
947 |
|
AK163759.1 |
- |
7, 8 |
836 |
|
AK164570.1 |
- |
7, 8 |
836 |
|
AK165173.1 |
- |
7, 8 |
847 |
|
AK170991.1 |
- |
7, 8 |
836 |
|
AK183348.1 |
- |
1, 2 |
61 |
|
AK205085.1 |
- |
3, 4 |
282 |
|
BC100452.1 |
- |
7, 8 |
840 |
6532792 |
View Full Product Details