Gene Transcripts

Gene Symbol
Ccnl2
Entrez Gene ID
56036
Gene Name
cyclin L2
Gene Aliases
1700010A01Rik, 1810019L15Rik, 2010319M22Rik, AA409936, AW261738, Ania-6b, HCLA-150, Pcee, SB138
Chromosome Location
Chr. 4: 155812084 - 155824543 on Build GRCm38
UniGene ID
Mm.23492
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207678.1 NP_997561.1 3 461
GenBank mRNA AB041605.1 - 3 438
AF211859.1 - 3 422
AK007552.1 - 3 458
AK008585.1 - 3 250
AK044204.1 - 3 398
AK048244.1 - 3 451
AK138938.1 - 3 172
AK156037.1 - 3 444
AK169857.1 - 3 437
AY337018.1 - 3 444
BC023747.1 - 3 436
BC132295.1 - 3 453 40130572
CD348864.1 - 3 461

View Full Product Details