Gene Transcripts

Gene Symbol
Pvrl1
Entrez Gene ID
58235
Gene Name
poliovirus receptor-related 1
Gene Aliases
AI835281, AW549174, Cd111, HIgR, HveC, PRR, PRR1, nectin-1
Chromosome Location
Chr. 9: 43744576 - 43807461 on Build GRCm38
UniGene ID
Mm.335096
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021424.2 NP_067399.2 2 421
GenBank mRNA AF239762.1 - 2 353
AF270977.1 - 2 353
AF297665.1 - 2 368
AK147479.1 - 1013
BC060694.1 - 2 421 6405088

View Full Product Details