Gene Transcripts

Gene Symbol
Tmub1
Entrez Gene ID
64295
Gene Name
transmembrane and ubiquitin-like domain containing 1
Gene Aliases
2010004O20Rik, AB030183, Hops
Chromosome Location
Chr. 5: 24445463 - 24448331 on Build GRCm38
UniGene ID
Mm.150098
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022418.3 NP_071863.1 2 491
GenBank mRNA AB030183.1 - 2 453
AK008109.1 - 2 425
AY603379.1 - 2 422
BC019547.1 - 2 447 4164765

View Full Product Details