Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025853.3 | NP_080129.2 | 9 | 892 | |
GenBank mRNA | AK006246.1 | - | 9 | 893 | |
AK017740.1 | - | 9 | 893 | ||
AK017813.1 | - | 9 | 847 | ||
AK139887.1 | - | 9 | 1462 | ||
AK145373.1 | - | 10 | 720 | ||
BC025079.1 | - | 9 | 875 | 3499844 | |
BC046807.1 | - | 9 | 853 | 6829342 |