Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025980.2 | NP_080256.2 | 1 | 466 | |
GenBank mRNA | AK012426.1 | - | 1 | 201 | |
AY046077.1 | - | 1 | 466 | ||
BC020293.1 | - | 1 | 463 | ||
BC048088.1 | - | 1 | 443 | ||
BC069891.1 | - | 1 | 219 | 5391567 |