Gene Transcripts

Gene Symbol
Iqcf4
Entrez Gene ID
67320
Gene Name
IQ motif containing F4
Gene Aliases
1700042N06Rik, mtIQ1
Chromosome Location
Chr. 9: 106568319 - 106570967 on Build GRCm38
UniGene ID
Mm.335431
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026090.2 NP_080366.2 3 473
GenBank mRNA BC048652.1 - 3 478
BC061014.1 - 3 473 6770556
DQ153246.1 - 3 502

View Full Product Details