Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001114087.2 | NP_001107559.1 | 3 | 247 | |
NM_001114088.2 | NP_001107560.1 | 3 | 247 | ||
NM_026131.4 | NP_080407.3 | 3 | 247 | ||
GenBank mRNA | AK003019.1 | - | 3 | 204 | |
AK003338.1 | - | 3 | 238 | ||
AK010141.1 | - | 3 | 236 | ||
AK010339.1 | - | 3 | 238 | ||
AK033091.1 | - | 239 | |||
AK049902.1 | - | 3 | 239 | ||
AK077998.1 | - | 3 | 239 | ||
AK128987.1 | - | 3 | 235 | ||
AK142933.1 | - | 3 | 230 | ||
AK167476.1 | - | 3 | 239 | ||
BC049565.1 | - | 3 | 235 | 6588160 | |
BC052698.1 | - | 3 | 218 | ||
BY056634.1 | - | 3 | 240 |