Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026191.2 | NP_080467.3 | 3 | 467 | |
GenBank mRNA | AK004610.1 | - | 3 | 431 | |
AK010512.1 | - | 3 | 447 | ||
AK029935.1 | - | 3 | 440 | ||
AK031000.1 | - | 3 | 414 | ||
AK135592.1 | - | 3 | 403 | ||
BC058276.1 | - | 3 | 385 | 6515479 |