Gene Transcripts

Gene Symbol
Otud3
Entrez Gene ID
73162
Gene Name
OTU domain containing 3
Gene Aliases
3110030K17Rik
Chromosome Location
Chr. 4: 138891063 - 138913947 on Build GRCm38
UniGene ID
Mm.238081
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028453.1 NP_082729.1 2 416
GenBank mRNA AK014098.1 - 2 458

View Full Product Details