Gene Transcripts

Gene Symbol
Syvn1
Entrez Gene ID
74126
Gene Name
synovial apoptosis inhibitor 1, synoviolin
Gene Aliases
1200010C09Rik, AW211966, C85322, D530017H19Rik, Hrd1
Chromosome Location
Chr. 19: 6045030 - 6053718 on Build GRCm38
UniGene ID
Mm.149870
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164709.1 NP_001158181.1 15, 16 1942
NM_028769.5 NP_083045.4 15, 16 2309
GenBank mRNA AK004688.2 - 15, 16 1796
AK122558.1 - 12 2763
AK143720.1 - 15, 16 1823
AK172097.1 - 10 2939
BC032931.1 - 4, 5 595
BC042199.1 - 15, 16 1804 4953165
BC046829.1 - 15, 16 1942 6811574
BC057917.1 - 15, 16 2223 5136398
BC080722.1 - 15, 16 1827 5323695

View Full Product Details