Gene Transcripts

Gene Symbol
Zbtb4
Entrez Gene ID
75580
Gene Name
zinc finger and BTB domain containing 4
Gene Aliases
2310026P19Rik, 9230111I22Rik, mKIAA1538
Chromosome Location
Chr. 11: 69765912 - 69784026 on Build GRCm38
UniGene ID
Mm.426361
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029348.2 NP_083624.2 4 1306
GenBank mRNA AK147212.1 - 4 1260
AK163593.1 - 4 1369
AK173196.1 - 1 1642

View Full Product Details