Gene Transcripts
- Gene Symbol
- Kcnip4
- Entrez Gene ID
- 80334
- Gene Name
- Kv channel interacting protein 4
- Gene Aliases
- AV032399, Calp, Calp250, KChIP4, KChIP4a
- Chromosome Location
- Chr. 5: 48389503 - 49525122 on Build GRCm38
- UniGene ID
- Mm.160172
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199242.1 |
NP_001186171.1
|
5, 6 |
641 |
|
NM_001199243.1 |
NP_001186172.1
|
4, 5 |
539 |
|
NM_001199244.1 |
NP_001186173.1
|
4, 5 |
682 |
|
NM_001199245.1 |
NP_001186174.1
|
4, 5 |
535 |
|
NM_030265.3 |
NP_084541.3
|
4, 5 |
467 |
|
GenBank mRNA |
AF305071.1 |
- |
4, 5 |
438 |
|
AF453243.1 |
- |
4, 5 |
375 |
|
AK039048.1 |
- |
4, 5 |
468 |
|
AK049662.1 |
- |
4, 5 |
419 |
|
AK133416.1 |
- |
4, 5 |
387 |
|
AK148685.1 |
- |
4, 5 |
684 |
|
AK165773.1 |
- |
4, 5 |
409 |
|
AY647240.1 |
- |
4, 5 |
418 |
|
BC051130.1 |
- |
5, 6 |
641 |
6532082 |
DQ148501.1 |
- |
5, 6 |
641 |
|
DQ148502.1 |
- |
3, 4 |
399 |
|
DQ148503.1 |
- |
4, 5 |
419 |
|
DQ148504.1 |
- |
4, 5 |
535 |
|
View Full Product Details