Gene Transcripts

Gene Symbol
Clec2h
Entrez Gene ID
94071
Gene Name
C-type lectin domain family 2, member h
Gene Aliases
Clrf, clr-f
Chromosome Location
Chr. 6: 128662385 - 128677374 on Build GRCm38
UniGene ID
Mm.197689
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053165.5 NP_444395.2 3 426
GenBank mRNA AF350410.1 - 3 405
AK017207.1 - 3 422
AK090364.1 - 3 426
AK136426.1 - 4 681
BC021766.1 - 3 402 4238060
HQ713437.1 - 3 333
HQ875061.1 - 3 369

View Full Product Details