Gene Transcripts

Gene Symbol
Trim34a
Entrez Gene ID
94094
Gene Name
tripartite motif-containing 34A
Gene Aliases
Trim34, Trim34-1
Chromosome Location
Chr. 7: 104244437 - 104262239 on Build GRCm38
UniGene ID
Mm.326945
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030684.3 NP_109609.2 4 1055
GenBank mRNA AF220139.1 - 4 952
AF220140.1 - 955

Gene Symbol
Trim34b
Entrez Gene ID
434218
Gene Name
tripartite motif-containing 34B
Gene Aliases
Gm15134, OTTMUSG00000019188, Trim34-2
Chromosome Location
Chr. 7: 104329471 - 104336617 on Build GRCm38
UniGene ID
Mm.485757
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243916.1 NP_001230845.1 3 796

View Full Product Details