Gene Transcripts

Gene Symbol
Ubqln4
Entrez Gene ID
94232
Gene Name
ubiquilin 4
Gene Aliases
A1Up, A1u, AI663987, CIP75, UBIN
Chromosome Location
Chr. 3: 88553677 - 88569725 on Build GRCm38
UniGene ID
Mm.303059
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033526.2 NP_277068.1 1, 2 187
GenBank mRNA AB040050.1 - 1, 2 187
AK150211.1 - 1, 2 125
AK156515.1 - 1, 2 140
AK160578.1 - 1, 2 139
BC017686.1 - 1, 2 149 4236887

View Full Product Details