Gene Transcripts

Gene Symbol
Eps8l3
Entrez Gene ID
99662
Gene Name
EPS8-like 3
Gene Aliases
AI504489, Eps8r3
Chromosome Location
Chr. 3: 107873860 - 107892900 on Build GRCm38
UniGene ID
Mm.108491
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133867.2 NP_598628.1 8 800
GenBank mRNA AK136584.1 - 8 797
AK167630.1 - 8 787
AY074932.1 - 8 816
BC014734.1 - 8 788 4218079

View Full Product Details