Gene Transcripts

Gene Symbol
Ptgr1
Entrez Gene ID
192227
Gene Name
prostaglandin reductase 1
Gene Aliases
Dig1, Ltb4dh
Chromosome Location
Chr. 5: 76110753 - 76129361 on Build Rnor_6.0
UniGene ID
Rn.10656
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138863.2 NP_620218.1 9 885
GenBank mRNA BC089775.1 - 9 885
FQ220898.1 - 9 906
U66322.2 - 8 850

View Full Product Details