Gene Transcripts

Gene Symbol
Whamm
Entrez Gene ID
293057
Gene Name
WAS protein homolog associated with actin, golgi membranes and microtubules
Gene Aliases
RGD1563506, Whdc1
Chromosome Location
Chr. 1: 143401986 - 143430356 on Build Rnor_6.0
UniGene ID
Rn.198506
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130728.1 NP_001124200.1 2 664

View Full Product Details