Gene Transcripts

Gene Symbol
Olr78
Entrez Gene ID
293221
Gene Name
olfactory receptor 78
Gene Aliases
ratchr1-160945784-160946731_ORF
Chromosome Location
Chr. 1: 168237950 - 168238897 on Build Rnor_6.0
UniGene ID
Rn.141908
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000135.1 NP_001000135.1 1 41

View Full Product Details