Gene Transcripts

Gene Symbol
Pla2g2f
Entrez Gene ID
690388
Gene Name
phospholipase A2, group IIF
Gene Aliases
Chromosome Location
Chr. 5: 157192047 - 157200167 on Build Rnor_6.0
UniGene ID
Rn.144878
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109587.1 NP_001103057.1 1, 2 246
XM_008764289.1 XP_008762511.1 2, 3 671

View Full Product Details