Gene Transcripts

Gene Symbol
LOC100911549
Entrez Gene ID
100911549
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase family member 6-like
Gene Aliases
Chromosome Location
Chr. 10: 192931 - 199426 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008767456.1 XP_008765678.1 3 405

Gene Symbol
Enpp6
Entrez Gene ID
306460
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 6
Gene Aliases
E-NPP 6
Chromosome Location
Chr. 16: 48315500 - 48437260 on Build Rnor_6.0
UniGene ID
Rn.8484
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107311.1 NP_001100781.1 4 648
XM_006253156.2 XP_006253218.1 4 666
XM_008771259.1 XP_008769481.1 4 666
GenBank mRNA BC158772.1 - 4 628

View Full Product Details