Gene Transcripts

Gene Symbol
Unc5d
Entrez Gene ID
306534
Gene Name
unc-5 homolog D (C. elegans)
Gene Aliases
Chromosome Location
Chr. 16: 67048092 - 67601002 on Build Rnor_6.0
UniGene ID
Rn.231130
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107319.2 NP_001100789.2 14 2023
XM_008771325.1 XP_008769547.1 11 1516

View Full Product Details