Gene Transcripts

Gene Symbol
Tigd3
Entrez Gene ID
309174
Gene Name
tigger transposable element derived 3
Gene Aliases
Chromosome Location
Chr. 1: 221248546 - 221252080 on Build Rnor_6.0
UniGene ID
Rn.49556
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107573.1 NP_001101043.1 2 654
XM_006230805.2 XP_006230867.1 2 1408

View Full Product Details