Gene Transcripts

Gene Symbol
Arhgap33
Entrez Gene ID
100362311
Gene Name
Rho GTPase activating protein 33
Gene Aliases
Chromosome Location
Chr. 1: 88977460 - 88990778 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008759253.1 XP_008757475.1 8 972

View Full Product Details