Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173315.1 | NP_775437.1 | 4 | 1498 | |
XM_008758879.1 | XP_008757101.1 | 4 | 1499 | ||
GenBank mRNA | AF016179.1 | - | 4 | 1498 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_003748744.2 | XP_003748792.2 | 1498 |