Gene Transcripts

Gene Symbol
Hbz
Entrez Gene ID
287168
Gene Name
hemoglobin, zeta
Gene Aliases
RGD1307486
Chromosome Location
Chr. 10: 15609348 - 15610826 on Build Rnor_6.0
UniGene ID
Rn.2703
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172845.1 NP_001166316.1 1 79
GenBank mRNA FQ221080.1 - 1 87
FQ221398.1 - 1 87
FQ222010.1 - 1 90

View Full Product Details