Gene Transcripts

Gene Symbol
Skp1
Entrez Gene ID
287280
Gene Name
S-phase kinase-associated protein 1
Gene Aliases
Skp1a
Chromosome Location
Chr. 10: 37594578 - 37609653 on Build Rnor_6.0
UniGene ID
Rn.3477
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007608.2 NP_001007609.1 4 369
XM_006246244.2 XP_006246306.1 4 668
GenBank mRNA BC058152.1 - 4 372
FQ210698.1 - 3 267
FQ212528.1 - 3 267
FQ212745.1 - 3 267
FQ212916.1 - 3 267
FQ213991.1 - 3 267
FQ214002.1 - 3 267
FQ214005.1 - 3 267
FQ214369.1 - 3 267
FQ214470.1 - 3 267
FQ214982.1 - 3 267
FQ215334.1 - 3 267
FQ215452.1 - 3 267
FQ219088.1 - 3 267
FQ219401.1 - 3 267
FQ224225.1 - 3 267
FQ224935.1 - 3 267
FQ229076.1 - 3 267
FQ229788.1 - 267
FQ230709.1 - 2 156
FQ230985.1 - 3 267

View Full Product Details