Gene Transcripts

Gene Symbol
V1ra14
Entrez Gene ID
297442
Gene Name
vomeronasal 1 receptor A14
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.215804
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008898.1 NP_001008898.1 1 619
GenBank mRNA AY510301.1 - 1 619

View Full Product Details